Tulobuterol Hydrochloride, 98%
Tulobuterol Hydrochloride, 98%
Tulobuterol Hydrochloride, 98%
Tulobuterol Hydrochloride, 98%
Thermo Scientific Chemicals
Tulobuterol Hydrochloride, 98%
beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl
Have Questions?
Catalog NumberQuantity
ALFJ61448.031 g
Catalog number ALFJ61448.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Quantity1 g
DescriptionWhite
For Use With (Application)beta-2-adrenergic receptor agonist
Recommended StorageKeep cold
Assay98%
View more
beta-2-adrenergic receptor agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C