Clindamycin hydrochloride monohydrate
Clindamycin hydrochloride monohydrate
Clindamycin hydrochloride monohydrate
Clindamycin hydrochloride monohydrate
Thermo Scientific Chemicals
Clindamycin hydrochloride monohydrate
Clindamycin hydrochloride, CAS # 21462-39-5, is the salt of clindamycin, a derivative of the broad-spectrum bacteriostatic antibiotic lincomycin. | CAS: 58207-19-5 | C18H34Cl2N2O5S | 461.439 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61409.031 g
Catalog number ALFJ61409.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialClindamycin hydrochloride monohydrate
TypeClindamycin Hydrochloride Monohydrate
Physical FormPowder
CAS58207-19-5
Health Hazard 1H315-H319-H335
View more
Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex
• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction
Application
• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well
• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies
RUO – Research Use Only
General References:
  1. FA El-Yazbi.; SM Blaih. Spectrophotometric and titrimetric determination of clindamycin hydrochloride in pharmaceutical preparations. Analyst. 1993, 118, (5), 577-579.
  2. K Ravikumar, B Sridhar. Clindamycin hydrochloride monohydrate and its ethanol solvate. Acta Cryst. 2010, C66, 97-100.