Leu-Enkephalin
Leu-Enkephalin
Leu-Enkephalin
Thermo Scientific Chemicals
Leu-Enkephalin
CAS: 81678-16-2615.684 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61408.MB25 mg
Catalog number ALFJ61408.MB
Price (MYR)
705.00
EA
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (MYR)
705.00
EA
Add to cart
Specifications
CAS81678-16-2
ColorWhite
FormatLyophilized Powder
Molecular Weight (g/mol)615.684
Product TypeLeu-Enkephalin
View more
Endogenous opioid neurotransmitter/neuromodulator that is an agonist at ? and δ opioid receptors, localization in brain parallels the localization of δ receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Endogenous opioid neurotransmittereuromodulator that is an agonist at μ and δ opioid receptors, localization in brain parallels the localization of δ receptors.
Solubility
Soluble in water (1 mg/ml ).
RUO – Research Use Only
General References:
  1. J. Alumets.; R. Hakanson.; F. Sundler.; K.-J. Chang. Leu-enkephalin-like material in nerves and enterochromaffin cells in the gut an immunohistochemical study. Histochemistry. 1978, 56, (3), 187-196.
  2. E. Cutz.; W. Chan.; N. S. Tracka. Bombesin, calcitonin and leu-enkephalin immunoreactivity in endocrine cells of human lung. Experiential. 1981, 37, (7), 765-767.
Contents & Storage
Store at -20°C