Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ61384.K21 L
Catalog number ALFJ61384.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBrilliant Blue R soln.
Name NoteReady-to-Use
CAS6104-59-2
Health Hazard 1H500
Health Hazard 2GHS H Statement
H311-H331-H370-H226
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Flammable liquid and vapor.