Polyvinylpyrrolidone, M.W. 360,000
Polyvinylpyrrolidone, M.W. 360,000
Polyvinylpyrrolidone, M.W. 360,000
Polyvinylpyrrolidone, M.W. 360,000
Thermo Scientific Chemicals
Polyvinylpyrrolidone, M.W. 360,000
A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
Catalog number ALFJ61381.A1
Price (MYR)
1,414.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,414.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyvinylpyrrolidone
ColorWhite
CAS9003-39-8
Physical FormPowder
Health Hazard 1H500
View more
A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.