Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.
Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.
Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.
Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.
Thermo Scientific Chemicals
Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.
A protein precipitant used to purify proteins | CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Catalog number ALFJ61270.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethyleneimine
ColorColorless to Yellow
CAS9002-98-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays, and to pretreat filters to increase the binding of proteins. Polyethyleneimine can also complex heavy metal ions. An effective permeabilizer of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on Polyethyleneimine cellulose-coated plates. It is a water soluble polymer used as a membrane lubricant, useful in filtering frozen cell membranes.