Dextran, MW ca 20,000
Dextran, MW ca 20,000
Dextran, MW ca 20,000
Dextran, MW ca 20,000
Thermo Scientific Chemicals
Dextran, MW ca 20,000
CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFJ61216.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDextran
ColorWhite
CAS9004-54-0
Physical FormPowder
Recommended StorageAmbient temperatures
View more
Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. Higher molecular weight dextrans have differing viscosities, aggregation and permeation properties.