Nigrosin, alcohol soluble
Nigrosin, alcohol soluble
Nigrosin, alcohol soluble
Nigrosin, alcohol soluble
Thermo Scientific Chemicals
Nigrosin, alcohol soluble
CAS: 11099-03-9 | C8H19ClN2O | 194.703 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61186.22100 g
Catalog number ALFJ61186.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNigrosin
CAS11099-03-9
Recommended StorageAmbient temperatures
RTECS NumberGE5800000
TSCAYes