Pectin Citrus
Thermo Scientific Chemicals
Pectin Citrus
CAS: 9000-69-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFJ61021.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPectin Citrus
CAS9000-69-5
ColorBrown to Yellow
Recommended StorageAmbient temperatures
RTECS NumberRX4280000
View more
Pectin Citrus is promoted in dietary supplement form as an alternative cancer treatment. It can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(RUO – Research Use Only.)  Promoted in dietary supplement form as an alternative cancer treatment, Pectin Citrus can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Jesper Harholt et. Al. Biosynthesis of pectin. Plant Physiology. 2010, 153,(2), 384-395.
  2. Robert Palin and Anja Geitmann. The role of pectin in plant morphogenesis. Biosystems. 2012, 109,(3), 397-402.