HEPES, 1.0M buffer soln., pH 6.5
HEPES, 1.0M buffer soln., pH 6.5
HEPES, 1.0M buffer soln., pH 6.5
HEPES, 1.0M buffer soln., pH 6.5
Thermo Scientific Chemicals
HEPES, 1.0M buffer soln., pH 6.5
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61017.AP500 mL
Catalog number ALFJ61017.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHEPES buffer soln.
CAS7365-45-9
Recommended StorageAmbient temperatures
Concentration1.0 M
TSCAYes