Rifampin, Molecular Biology Reagent
Rifampin, Molecular Biology Reagent
Rifampin, Molecular Biology Reagent
Rifampin, Molecular Biology Reagent
Thermo Scientific Chemicals
Rifampin, Molecular Biology Reagent
Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe. | CAS: 13292-46-1 | C43H58N4O12 | 822.953 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFJ60836.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRifampin
TypeRifampicin
Physical FormPowder
CAS13292-46-1
Health Hazard 1H302-H315-H319-H335
View more
Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifampin is an antibiotic used in the treatment of tuberculosis and its related microbacterial infections. It is involved in the inhibition of DNA polymerase and RNA polymerase. As an antipruritic agent, it is used in the autoimmune cholestatic liver disease and primary biliary cirrhosis (PBC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica
Applications
  • Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria
  • It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis
RUO – Research Use Only
General References:
  1. Tarakji, K.G.; Mittal, S.; Kennergren, C.; Corey, R.; Poole, J.E.; Schloss, E.; Gallastegui, J.; Pickett, R.A.; Evonich, R.; Philippon, F.; McComb, J.M., Roark, S.F.; Sorrentino, D.; Sholevar, D.; Cronin, E.; Berman, B., Riggio, D.; Biffi, M.; Khan, H.; Silver, M.T.; Collier, J.; Eldadah, Z.; Wright, D.J.; Lande, J.D.; Lexcen, D.R.; Cheng, A.; Wilkoff, B.L. WRAP-IT Investigators. Antibacterial Envelope to Prevent Cardiac Implantable Device Infection. N Engl J Med. 2019, 380(20), 1895-1905.
  2. Swindells, S.; Ramchandani, R.; Gupta, A.; Benson, C.A.; Leon-Cruz, J.; Mwelase, N.; Jean Juste, M.A.; Lama, J.R.; Valencia, J.; Omoz-Oarhe, A.; Supparatpinyo, K.; Masheto, G.; Mohapi L, da Silva Escada, R.O.; Mawlana, S.; Banda, P.; Severe, P.; Hakim, J.; Kanyama, C.; Langat, D.; Moran, L.; Andersen, J.; Fletcher, C.V.; Nuermberger, E.; Chaisson, R.E., BRIEF TB/A5279 Study Team. One Month of Rifapentine plus Isoniazid to Prevent HIV-Related Tuberculosis. N Engl J Med. 2019, 380(11), 1001-1011.