1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer
Thermo Scientific Chemicals
1,3-Dihydroxyacetone dimer
CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60592.065 g
Catalog number ALFJ60592.06
Price (MYR)
302.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
302.00
EA
Add to cart
Specifications
Chemical Name or Material1,3-Dihydroxyacetone dimer
Melting Point75°C to 80°C
CAS62147-49-3
ColorWhite
Recommended StorageKeep cold
View more
1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.
Solubili
RUO – Research Use Only
General References:
  1. Duncan Gardiner. The dimeric forms of some α-hydroxycarbonyl compounds. Carbohydr. Res. 1966, 2 (3),234-239 .
  2. Varoujan A. Yaylayan.;Susan Harty-Majors.; Ashraf A. Ismail. Investigation of dl-glyceraldehyde-dihydroxyacetone interconversion by FTIR spectroscopy.Carbohydr. Res. 1999, 318 (1-4),20-25 .