Naltrexone hydrochloride, Thermo Scientific Chemicals
Naltrexone hydrochloride, Thermo Scientific Chemicals
Naltrexone hydrochloride, Thermo Scientific Chemicals
Naltrexone hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naltrexone hydrochloride, Thermo Scientific Chemicals
A congener of naloxone and also an opioid antagonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFJ60590.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialNaltrexone hydrochloride
CAS16676-29-2
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.
Solubility
Soluble in water at 50mg/ml.
Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.
RUO – Research Use Only
General References:
  1. Abdelkader, H.; Pierscionek, B.; Alany, R. G. Novel in situ gelling ocular films for the opioid growth factor-receptor antagonist-naltrexone hydrochloride: fabrication, mechanical properties, mucoadhesion, tolerability and stability studies. Int. J. Pharm. 2014, 477 (1-2), 631-642.
  2. Darwish, M.; Bond, M.; Yang, R.; Tracewell, W.; Robertson J, P. Assessment of Alcohol-Induced Dose Dumping with a Hydrocodone Bitartrate Extended-Release Tablet Formulated with CIMA Abuse Deterrence Technology. Clin. Drug Invest. 2015, 35 (10), 645-652.