Phosphate, 0.2M buffer soln., pH 4.4
Phosphate, 0.2M buffer soln., pH 4.4
Phosphate, 0.2M buffer soln., pH 4.4
Phosphate, 0.2M buffer soln., pH 4.4
Thermo Scientific Chemicals
Phosphate, 0.2M buffer soln., pH 4.4
Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. | CAS: 7558-80-7119.976 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60467.K21 L
Catalog number ALFJ60467.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Concentration0.2M buffer soln.
Molecular Weight (g/mol)119.976
Quantity1 L
PackagingPlastic Bottle
CAS7558-80-7
View more
This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Keep cold