Hexamethonium bromide, 98+%
Hexamethonium bromide, 98+%
Hexamethonium bromide, 98+%
Hexamethonium bromide, 98+%
Thermo Scientific Chemicals
Hexamethonium bromide, 98+%
Nicotinic acetyl choline receptor antagonist | CAS: 55-97-0 | C12H30Br2N2 | 362.194 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60466.1425 g
Catalog number ALFJ60466.14
Price (MYR)
537.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
537.00
EA
Add to cart
Specifications
Chemical Name or MaterialHexamethonium bromide
CAS55-97-0
Melting Point282°C to 285°C (decomposition)
ColorWhite
Recommended StorageAmbient temperatures
View more
Nicotinic acetyl choline receptor antagonist