Chlorogenic acid
Chlorogenic acid
Chlorogenic acid
Thermo Scientific Chemicals
Chlorogenic acid
An analog of caffeic acid | CAS: 327-97-9 | C16H18O9 | 354.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFJ60457.06
Price (MYR)
2,077.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,077.00
EA
Add to cart
Specifications
Chemical Name or MaterialChlorogenic acid
CAS327-97-9
Melting Point210°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberGU8480000
View more
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
Solubility
Soluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kamiyama, M.; Moon, J.; Jang, H. W.; Shibamoto, T. Role of Degradation Products of Chlrogenic Acid in the Antioxidant Activity of Roasted Coffee. J. Agric. Food Chem. 2015, 63 (7), 1996-2005.
  2. Peng, H.; Kather, M.; Rubsam, K.; Jakob, F.; Schwanebwerg, U.; Pich, A. Chlorogenic Acid Biosynthesis Appears Linked with Suberin Production in Potato Tuber (Solanum tuberosum). J. Agric. Food Chem. 2015, 63 (19), 4902-4913.