Theophylline, Thermo Scientific Chemicals
Theophylline, Thermo Scientific Chemicals
Theophylline, Thermo Scientific Chemicals
Theophylline, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Theophylline, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ60203.5H454 g
Catalog number ALFJ60203.5H
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
454 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTheophylline
CAS58-55-9
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Theophylline has been used to study its effects on rat gene expression in the ubiquitin-proteasome pathway that regulates spermatogenesis and epididymal sperm quality. Theophylline has also been used as an internal standard for the measurement of plasma paracetamol levels in humans.
Solubility
Soluble in ethanol (moderately), DMSO, alkali hydroxides, ammonia, dilute hyrochloric acid, nitric acid, dilute aqueous acid, and dilute aqueous base. Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Mark W Tengowski; Dongyan Feng; Miriam Sutovsky; Peter Sutovsky. Differential expression of genes encoding constitutive and inducible 20S proteasomal core subunits in the testis and epididymis of theophylline- or 1,3-dinitrobenzene-exposed rats. Biology of Reproduction, 2007, 76(1), 149-163.
  2. F J Sevilla-Tirado et. al.Bioavailability of two new formulations of paracetamol, compared with three marketed formulations, in healthy volunteers.. Methods and Findings in Experimental and Clinical Pharmacology, 2003,25,(7), 531-535.