Separating or resolving buffer (4X)
Separating or resolving buffer (4X)
Thermo Scientific Chemicals
Separating or resolving buffer (4X)
Have Questions?
Catalog NumberQuantity
ALFJ60181.K21L
Catalog number ALFJ60181.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1L
Request bulk or custom format
Specifications
Concentration4X
FormLiquid
Quantity1L
Storage RequirementsAmbient temperatures