5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
Substrate for beta-galactosidase | CAS: 107021-38-5 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60151.MD250 mg
Catalog number ALFJ60151.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
CAS107021-38-5
ColorWhite
Recommended StorageStore at -20°C
TSCANo
View more
Substrate for beta-galactosidase