Chlortetracycline hydrochloride
Chlortetracycline hydrochloride
Chlortetracycline hydrochloride
Chlortetracycline hydrochloride
Thermo Scientific Chemicals
Chlortetracycline hydrochloride
Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria | CAS: 64-72-2 | C22H24Cl2N2O8 | 515.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60095.1425 g
Catalog number ALFJ60095.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChlortetracycline hydrochloride
CAS64-72-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.
Solubility
Soluble in water
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Lindsay Gillan; Gareth Evans; W M C Maxwell. Flow cytometric evaluation of sperm parameters in relation to fertility potential. Theriogenology.200563(2), 445-457.
  2. Y H Choi; Y Toyodal. Cyclodextrin removes cholesterol from mouse sperm and induces capacitation in a protein-free medium. Biology of Reproduction.199859(6), 1328-1333.