D-(+)-Glucose, 1M aq. soln., sterile-filtered
D-(+)-Glucose, 1M aq. soln., sterile-filtered
D-(+)-Glucose, 1M aq. soln., sterile-filtered
D-(+)-Glucose, 1M aq. soln., sterile-filtered
Thermo Scientific Chemicals
D-(+)-Glucose, 1M aq. soln., sterile-filtered
CAS: 50-99-7 | C6H12O6 | 180.156 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60067.AK250 mL
Catalog number ALFJ60067.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialD-(+)-Glucose
Name Notesterile
CAS50-99-7
Recommended StorageAmbient temperatures
TSCAYes
View more
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.
Solubility
Not miscible or difficult to mix with water. Soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Aoi Ono; Yoshio Masuda. Separation behaviour of some isomeric organic compounds on sugars, sugar alcohols and their mixed phases by gas-liquid chromatography. Journal of Chromatography A, 1980, 197(2), 251-257.
  2. G Thierbach; and H Reichenbach. Myxothiazol, a new antibiotic interfering with respiration. Cancer Science, 1981,19(4), 504-507.