Peroxidase, horseradish
Peroxidase, horseradish
Thermo Scientific Chemicals
Peroxidase, horseradish
Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number ALFJ60026.MC
Price (MYR)
875.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
875.00
EA
Add to cart
Specifications
Recommended StorageStore at -20°C
EINECS Number232-668-6
Quantity100 mg
FormLyophilized
Unit SizeEA
Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.