Paraformaldehyde Solution, 4% in PBS
Paraformaldehyde Solution, 4% in PBS
Thermo Scientific Chemicals
Paraformaldehyde Solution, 4% in PBS
Have Questions?
Catalog NumberQuantity
ALFJ19943.K21 L
Catalog number ALFJ19943.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde Solution
Physical FormSolution
Health Hazard 1H317-H318-H351
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
PackagingPlastic bottle