Check Order Status
Sodium Dodecyl Sulfate (SDS), ≥95%
Element Search
Structure Search
Thermo Scientific Chemicals
Sodium Dodecyl Sulfate (SDS), ≥95%
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ALFJ18220.36
500 g
Plastic bottle
Catalog number
ALFJ18220.36
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Dodecyl Sulfate (SDS)
CAS
151-21-3
Health Hazard 1
H302-H315-H318-H500
Physical Form
Crystalline Powder
Health Hazard 3
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c