Check Order Status
L-Glutamine, ≥98.5%, Ultrapure
Element Search
Structure Search
Thermo Scientific Chemicals
L-Glutamine, ≥98.5%, Ultrapure
CAS: 56-85-9 | C5H10N2O3 | 146.146 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ALFJ16285.36
500 g
Plastic Bottle
Catalog number
ALFJ16285.36
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
L-Glutamine
CAS
56-85-9
Health Hazard 1
H315-H319-H320-H335-H336
Health Hazard 3
P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage
Ambient temperatures