Diethyl Pyrocarbonate, >97%, Ultrapure
Diethyl Pyrocarbonate, >97%, Ultrapure
Diethyl Pyrocarbonate, >97%, Ultrapure
Diethyl Pyrocarbonate, >97%, Ultrapure
Thermo Scientific Chemicals
Diethyl Pyrocarbonate, >97%, Ultrapure
CAS: 1609-47-8 | C6H10O5 | 162.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Plastic bottle
Catalog number ALFJ14710.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl Pyrocarbonate
CAS1609-47-8
Health Hazard 1H227-H302-H315-H319-H332-H335
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
Melting Point69°C
View more
  • A strong, nonspecific inhibitor of ribonuclease
  • Reacts with the N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Strong, nonspecific inhibitor of ribonuclease
  • Reacts with N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer
RUO – Research Use Only