N-Boc-iminodiacetic acid, 98%
N-Boc-iminodiacetic acid, 98%
N-Boc-iminodiacetic acid, 98%
N-Boc-iminodiacetic acid, 98%
Thermo Scientific Chemicals
N-Boc-iminodiacetic acid, 98%
CAS: 56074-20-5 | C9H15NO6 | 233.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFH66868.1425 g
Catalog number ALFH66868.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Boc-iminodiacetic acid
CAS56074-20-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c