2-Amino-2'-chloro-5-nitrobenzophenone, 98%
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
Thermo Scientific Chemicals
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
CAS: 2011-66-7 | C13H9ClN2O3 | 276.68 g/mol
Have Questions?
Catalog NumberQuantity
ALFH66440.065 g
Catalog number ALFH66440.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-2'-chloro-5-nitrobenzophenone
CAS2011-66-7
Melting Point119°C to 123°C
Storage Note 12° to 8°C
ColorYellow