N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
CAS: 35141-36-7 | C9H24ClNO3Si | 257.83 g/mol
Have Questions?
Catalog NumberQuantity
ALFH66414.36500 g
Catalog number ALFH66414.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride
Name Notein 50% methanol
CAS35141-36-7
Health Hazard 1H225-H301+H311+H331-H370
Health Hazard 2GHS H Statement
H225-H311-H331-H370-H335-H302-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.