(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals
(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals
(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals
(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH66260.031 g
Catalog number ALFH66260.03
Price (MYR)
1,063.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,063.00
EA
Add to cart
Specifications
Chemical Name or Material(2-Oxo-1-pyrrolidinyl)acetic acid
CAS53934-76-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c