2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
Thermo Scientific Chemicals
2-Bromo-6-fluorobenzaldehyde, 98%
CAS: 360575-28-6 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFH64689.22
Price (MYR)
3,348.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
3,348.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromo-6-fluorobenzaldehyde
CAS360575-28-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Christian Ruzié; Jolanta Karpinska; Alan R. Kennedy; and Yves H. Geerts. Synthesis of 1,6-, 2,7-, 3,8-, and 4,9-Isomers of Didodecyl[1]benzothieno[3,2-b][1]benzothiophenes. J. Org. Chem. 2013 78(15), 7741-7748.
  2. Xinzhi Chen; Shaodong Zhou; Chao Qian; Chaohong He. Synthesis of 1H-indazole: a combination of experimental and theoretical studies. Research on Chemical Intermediates. 2013 38(7), 1619-1628.