3-Bromo-2-fluorobenzaldehyde, 97%
3-Bromo-2-fluorobenzaldehyde, 97%
3-Bromo-2-fluorobenzaldehyde, 97%
3-Bromo-2-fluorobenzaldehyde, 97%
Thermo Scientific Chemicals
3-Bromo-2-fluorobenzaldehyde, 97%
CAS: 149947-15-9 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH64650.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Bromo-2-fluorobenzaldehyde
CAS149947-15-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It can be used as pharmaceutical raw material and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Les A. Dakin; Michael H. Block; Huawei Chen; Erin Code; James E. Dowling; Xiaomei Feng; Andrew D. Ferguson; Isabelle Green; Alexander W. Hird; Tina Howard; Erika K. Keeton; Michelle L. Lamb; Paul D. Lyne; Hannah Pollard; Jon Read; Allan J. Wu; Tao Zhang; Xiaolan Zheng. Discovery of novel benzylidene-1,3-thiazolidine-2,4-diones as potent and selective inhibitors of the PIM-1, PIM-2, and PIM-3 protein kinases. Bioorganic & Medicinal Chemistry Letters. 2012 22(14), 4599-4604.