Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
Thermo Scientific Chemicals
Benzenesulfonic acid isopropyl ester, 95%
CAS: 6214-18-2 | C9H12O3S | 200.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFH64523.14
Price (MYR)
4,170.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
4,170.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzenesulfonic acid isopropyl ester
CAS6214-18-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
A sulfonate ester that acts as a potential effects in drug substances impurity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.
Solubility
Soluble in chloroform and ethyl acetate.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Guo T.; Shi, Y.; Zheng, L., et al. Rapid and simultaneous determination of sulfonate ester genotoxic impurities in drug substance by liquid chromatography coupled to tandem mass spectrometry: Comparison of different ionization modes. J Chromatogr A. 2014 135573-79.
  2. John N. Bonfigliol. Directed ortho-lithiation of alkyl arenesulfonates. J. Org. Chem. 1986 51(14), 2833-2835.