2-Bromo-3-fluorobenzaldehyde, 98%
2-Bromo-3-fluorobenzaldehyde, 98%
2-Bromo-3-fluorobenzaldehyde, 98%
2-Bromo-3-fluorobenzaldehyde, 98%
Thermo Scientific Chemicals
2-Bromo-3-fluorobenzaldehyde, 98%
CAS: 891180-59-9 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH64400.14
Price (MYR)
1,522.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,522.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromo-3-fluorobenzaldehyde
CAS891180-59-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c