3-Bromo-5-fluorobenzaldehyde, 95%
3-Bromo-5-fluorobenzaldehyde, 95%
3-Bromo-5-fluorobenzaldehyde, 95%
3-Bromo-5-fluorobenzaldehyde, 95%
Thermo Scientific Chemicals
3-Bromo-5-fluorobenzaldehyde, 95%
Form: solid | CAS: 188813-02-7 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH64042.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Bromo-5-fluorobenzaldehyde
CAS188813-02-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air , heat.
RUO – Research Use Only
General References:
  1. Gaifa Lai,; J. Robert Merritt,; Zhenmin He,; Daming Feng,; Jianhua Chao,; Michael F. Czarniecki,; Laura L. Rokosz.Synthesis and structure-activity relationships of new disubstituted phenyl-containing 3,4-diamino-3-cyclobutene-1,2-diones as CXCR2 receptor antagonists. Bioorganic & Medicinal Chemistry Letters. year of publication , 18 (16), 1864-1868.