1-Indoleacetic acid, 95%
1-Indoleacetic acid, 95%
1-Indoleacetic acid, 95%
1-Indoleacetic acid, 95%
Thermo Scientific Chemicals
1-Indoleacetic acid, 95%
CAS: 24297-59-4 | C10H9NO2 | 175.187 g/mol
Have Questions?
Catalog NumberQuantity
ALFH63935.MD250 mg
Catalog number ALFH63935.MD
Price (MYR)
684.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
684.00
EA
Add to cart
Specifications
Chemical Name or Material1-Indoleacetic acid
CAS24297-59-4
Health Hazard 1H302-H318
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c