5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
Thermo Scientific Chemicals
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
CAS: 253870-02-9 | C8H9NO3 | 167.164 g/mol
Have Questions?
Catalog NumberQuantity
ALFH63711.065 g
Catalog number ALFH63711.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid
CAS253870-02-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Kai Lv; Li-Li Wang; Ming-Liang Liu; Xin-Bo Zhou; Shi-Yong Fan; Hong-Ying Liu; Zhi-Bing Zheng; Song L. Synthesis and antitumor activity of 5-[1-(3-(dimethylamino)propyl)-5-halogenated-2-oxoindolin-(3Z)-ylidenemethyl]-2,4-dimethyl-1H-pyrrole-3-carboxamides. Bioorganic & Medicinal Chemistry Letters. 2011, 10(21), 3062-3065.