1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number ALFH62185.MD
Price (MYR)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material1-Boc-N-Fmoc-D-tryptophan
CAS163619-04-3
Recommended StorageKeep cold; Store under Nitrogen
TSCANo
Molecular FormulaC31H30N2O6