Anthraquinone-2-carboxylic acid, 98%
Anthraquinone-2-carboxylic acid, 98%
Anthraquinone-2-carboxylic acid, 98%
Anthraquinone-2-carboxylic acid, 98%
Thermo Scientific Chemicals
Anthraquinone-2-carboxylic acid, 98%
CAS: 117-78-2 | C15H8O4 | 252.225 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60929.031 g
Catalog number ALFH60929.03
Price (MYR)
419.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
419.00
EA
Add to cart
Specifications
Chemical Name or MaterialAnthraquinone-2-carboxylic acid
CAS117-78-2
Melting Point287°C to 289°C
Recommended StorageAmbient temperatures
EINECS Number204-207-9
View more
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.
Solubility
Soluble in hot Acetic acid(almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Navid Mogharrab,; Hedayatollah Ghourchian. Anthraquinone 2-carboxylic acid as an electron shuttling mediator and attached electron relay for horseradish peroxidase. Electrochemistry Communications. 2005, 7 (5), 466-471.
  2. Sang Woo Han,; Sang Woo Joo,; Tai Hwan Ha,; Yunsoo Kim,; Kwan Kim.Adsorption Characteristics of Anthraquinone-2-carboxylic Acid on Gold . J. Phys. Chem. B, year of publication , 104(50), 11987-11995.