4-Methylhippuric acid, 98%, Thermo Scientific™
4-Methylhippuric acid, 98%, Thermo Scientific™
4-Methylhippuric acid, 98%, Thermo Scientific™
4-Methylhippuric acid, 98%, Thermo Scientific™
4-Methylhippuric acid, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH60754.031 g
Catalog number ALFH60754.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methylhippuric acid
CAS27115-50-0
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point163°C to 165°C
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.