1,2-Benzisothiazol-3-one, 97%
1,2-Benzisothiazol-3-one, 97%
1,2-Benzisothiazol-3-one, 97%
1,2-Benzisothiazol-3-one, 97%
Thermo Scientific Chemicals
1,2-Benzisothiazol-3-one, 97%
CAS: 2634-33-5 | C7H5NOS | 151.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH60492.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Benzisothiazol-3-one
CAS2634-33-5
Health Hazard 1H302-H315-H317-H318
Health Hazard 2GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.
Health Hazard 3P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P501c
View more
It is an antimicrobial agent and a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an antimicrobial agent and a pharmaceutical intermediate.
Solubility
Soluble in dichloromethane, dimethyl sulfoxide, methanol.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Keith D. Combrink .; H. Belgin Gülgeze.; Nicholas A. Meanwell .; Bradley C. Pearce.; Pi Zulan .; Gregory S. Bisacchi.; Daniel G. M. Roberts.; Paul Stanley.; Steven M. Seiler. 1,2-Benzisothiazol-3-one 1,1-Dioxide Inhibitors of Human Mast Cell Tryptase. J. Med. Chem. 1998, 41 (24),4854-4860.
  2. Magid Abou-Gharbia.; John A. Moyer.; Usha Patel.; Michael Webb.; Guy Schiehser.; Terrance Andree.; J. Thomas Haskins. Synthesis and structure-activity relationship of substituted tetrahydro- and hexahydro-1,2-benzisothiazol-3-one 1,1-dioxides and thiadiazinones: potential anxiolytic agents. J. Med. Chem. 1989, 32 (5),1024-1033.