2,3,6-Trimethylphenol, 95%
2,3,6-Trimethylphenol, 95%
2,3,6-Trimethylphenol, 95%
2,3,6-Trimethylphenol, 95%
Thermo Scientific Chemicals
2,3,6-Trimethylphenol, 95%
CAS: 2416-94-6 | C9H12O | 136.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60489.1425 g
Catalog number ALFH60489.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,3,6-Trimethylphenol
CAS2416-94-6
Health Hazard 1H315-H317-H318
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
View more
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.
Solubility
Soluble in methanol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only
General References:
  1. O.A. Kholdeeva; A.V. Golovin; R.I. Maksimovskaya; I.V. Kozhevnikov. Oxidation of 2,3,6-trimethylphenol in the presence of molybdovanadophosphoric heteropoly acids. Journal of Molecular Catalysis.1992, 75 235-244.
  2. Katsuomi Takehira; Masao Shimizu; Yoshihito Watanabe; Hideo Orita; Takashi Hayakawa. A novel oxygenation of 2,3,6-trimethylphenol to trimethyl-p-benzoquinone by dioxygen with copper(II) chloride/amine hydrochloride catalyst. Tetrahedron Letters.1989, 30 6691-6692.