L-Dihydroorotic acid, 98%
L-Dihydroorotic acid, 98%
L-Dihydroorotic acid, 98%
L-Dihydroorotic acid, 98%
Thermo Scientific Chemicals
L-Dihydroorotic acid, 98%
CAS: 5988-19-2 | C5H6N2O4 | 158.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH60429.06
Price (MYR)
4,028.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
4,028.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Dihydroorotic acid
CAS5988-19-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
L-Dihydroorotic acid a metabolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Dihydroorotic acid a metabolite.
Solubility
Soluble in water (partly), and dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Charles S. Miller.; Janice T. Gordon.; Edward L. Engelhardt. Dihydroörotic Acid.J. Am. Chem. Soc.. 1953, 75 (23), 6086-6087.