Tetradecanedioic acid, 98%
Tetradecanedioic acid, 98%
Tetradecanedioic acid, 98%
Tetradecanedioic acid, 98%
Thermo Scientific Chemicals
Tetradecanedioic acid, 98%
CAS: 821-38-5 | C14H26O4 | 258.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH60254.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetradecanedioic acid
CAS821-38-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.
RUO – Research Use Only
General References:
  1. Masahiko Saitoh; Shizuo Fujisaki; Yasuhiro Ishii; Takeshi Nishiguchi. Convenient selective monoesterification of α, ω-dicarboxylic acids catalyzed by ion-exchange resins. Tetrahedron Letters. 1996, 37,(37), 6733-6736. 
  2. Maria Victoria Roux; Manuel Temprado; James S.Chickos. Vaporization, fusion and sublimation enthalpies of the dicarboxylic acids from C4 to C14 and C16. The Journal of Chemical Thermodynamics. 2005, 37,(9), 941-953.