Folinic acid calcium salt hydrate, 98%
Folinic acid calcium salt hydrate, 98%
Folinic acid calcium salt hydrate, 98%
Folinic acid calcium salt hydrate, 98%
Thermo Scientific Chemicals
Folinic acid calcium salt hydrate, 98%
CAS: 1492-18-8 | C20H21CaN7O7 | 511.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFH60181.MD
Price (MYR)
975.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (MYR)
975.00
EA
Add to cart
Specifications
Chemical Name or MaterialFolinic acid calcium salt hydrate
CAS1492-18-8
Health Hazard 1H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c