Terpineol, mixed isomers, 98%
Terpineol, mixed isomers, 98%
Terpineol, mixed isomers, 98%
Terpineol, mixed isomers, 98%
Thermo Scientific Chemicals
Terpineol, mixed isomers, 98%
CAS: 8000-41-7 | C10H18O | 154.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60146.AP500 mL
Catalog number ALFH60146.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTerpineol
Name Notemixed isomers
CAS8000-41-7
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
Solubility
Miscible with organic solvents. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Prakash, B.; Singh, P.; Goni, R.; Raina, A. K.; Dubey, N. K. Efficacy of essential oil, phenyl ethyl alcohol and alfa-terpineol against isolated molds from walnut and their antiaflatoxigenic and antioxidant activity. J. Food Sci. Technol. 2015, 52 (4), 2220-2228.
  2. Lee, S. Y.; Kim, S. H.; Hong, C. Y.; Park, S. Y.; Choi, I. G. Biotransformation of (-)-α-pinene and geraniol to alfa-terpineol and p-menthane-3, 8-diol by the white rot fungus, Polyporus brumalis. J. Microbiol. 2015, 53 (7), 462-467.