beta-Carotene, 99%
beta-Carotene, 99%
beta-Carotene, 99%
Thermo Scientific Chemicals
beta-Carotene, 99%
CAS: 7235-40-7 | C40H56 | 536.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH60106.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialbeta-Carotene
CAS7235-40-7
Melting Point178°C to 179°C
Storage Note 1-30° to -10°C
Recommended StorageStore at -20°C; Store under Argon
View more
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
Solubility
Soluble in hexane, dimethyl sulfoxide, benzene, chloroform, cyclohexane. Insoluble in water.
Notes
Air & Light Sensitive. Store in a cool place.
RUO – Research Use Only
R. Peto.; R. Doll.; J. D. Buckley.; M. B. Sporn. Can dietary beta-carotene materially reduce human cancer rates? Nature. 1981, 290201 - 208.