Oxcarbazepine, 98%
Oxcarbazepine, 98%
Oxcarbazepine, 98%
Thermo Scientific Chemicals
Oxcarbazepine, 98%
CAS: 28721-07-5 | C15H12N2O2 | 252.273 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60014.MD250 mg
Catalog number ALFH60014.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialOxcarbazepine
CAS28721-07-5
Recommended StorageAmbient temperatures
RTECS NumberHN8445000
EINECS Number249-188-8
View more
Oxcarbazepine is a sodium channel protein inhibitor. It is an anticonvulsant and mood-stab.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in DMSO, methanol, water, ethanol and acetone.
Notes
Store at -20°C.
RUO – Research Use Only
General References:
  1. Mogens Dam (Chairman).; Rolf Ekberg.; Yngve Løyning.; Olli Waltimo.; Kurt Jakobsen. A double-blind study comparing oxcarbazepine and carbamazepine in patients with newly diagnosed, previously untreated epilepsy. Epilepsy Research. 1989, 3 (1),70-76.
  2. M. L. Friis.;O. Kristensen.; J. Boas.; M. Dalby.; S. H. Deth.; L. Gram.; M. Mikkelsen.; B. Pedersen.; A. Sabers.; J. Worm-Petersen.; D. Andersen.; P. K. Jensen. Therapeutic experiences with 947 epileptic out-patients in oxcarbazepine treatment. Acta Neurologica Scandinavica. 1993, 87 (3),224-227.