4-Hydroxybenzenesulfonic acid, 65%
4-Hydroxybenzenesulfonic acid, 65%
4-Hydroxybenzenesulfonic acid, 65%
4-Hydroxybenzenesulfonic acid, 65%
Thermo Scientific Chemicals
4-Hydroxybenzenesulfonic acid, 65%
CAS: 98-67-9 | C6H6O4S | 174.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALFH59917.K2
Price (MYR)
1,370.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,370.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzenesulfonic acid
CAS98-67-9
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. R Campos; A Kandelbauer; K.H Robra; Artur Cavaco-Paulo; G.M Gübitz. Indigo degradation with purified laccases from Trametes hirsuta and Sclerotium rolfsii. Journal of Biotechnology,2001, 89 (2-3), 131-139.
  2. A M Lambeir; H B Dunford; M A Pickard. Kinetics of the oxidation of ascorbic acid, ferrocyanide and p-phenolsulfonic acid by chloroperoxidase compounds I and II. European Journal of Biochemistry. 1987, 163(1), 123-127.