1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™
1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™
1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™
1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™
1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH59803.065 g
Catalog number ALFH59803.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-n-Butyl-3-methylimidazolium nitrate
CAS179075-88-8
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.